CAS RN 301305-73-7
IUPAC Name:
2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Synonyms:
2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Molecular Weight (g/mol):
360.43
Molecular Formula:
C18H20N2O4S
InChi Key:
FSPQCTGGIANIJZ-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C=C(C=C1)C(=O)NC1=C(C(N)=O)C2=C(CCCC2)S1
1
–
3
of
3
results