accessibility menu, dialog, popup

UserName

CAS RN 3019-71-4

IUPAC Name: trichloroethanecarbonyl isocyanate
Synonyms: trichloroethanecarbonyl isocyanate
Molecular Weight (g/mol): 188.39
Molecular Formula: C3Cl3NO2
InChi Key: GRNOZCCBOFGDCL-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=O)N=C=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)