missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetyl isocyanate, 97%

Product Code. 11359953
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359953 50 g 50g
11332715 2 g 2g
11342715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359953 Supplier Thermo Scientific Alfa Aesar Supplier No. L00226.18
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3019-71-4 | C3Cl3NO2 | 188.39 g/mol

Trichloroacetyl isocyanate is used as an in-situ derivatizing reagent for characterization of alcohols, glycols, phenols, and amines by nuclear magnetic resonance. It is also used in anion chemical ionization mass spectrometry and in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. Further, it serves as a reagent for the conversion of alcohols into carbamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetyl isocyanate is used as an in-situ derivatizing reagent for characterization of alcohols, glycols, phenols, and amines by nuclear magnetic resonance. It is also used in anion chemical ionization mass spectrometry and in catalytic one-pot dehydrative glycosylation of 1-hydroxy carbohydrate. Further, it serves as a reagent for the conversion of alcohols into carbamates.

Solubility
Miscible with dichloromethane, ether, terahydrofuran and protic solvents.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3019-71-4
Molecular Formula C3Cl3NO2
Molecular Weight (g/mol) 188.39
MDL Number MFCD00002033
InChI Key GRNOZCCBOFGDCL-UHFFFAOYSA-N
Synonym trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
PubChem CID 76400
SMILES ClC(Cl)(Cl)C(=O)N=C=O

Specifications

Density 1.585
Boiling Point 58°C to 60°C (20 mmHg)
Flash Point 65°C (149°F)
Odor Acrid
Refractive Index 1.48
Quantity 50 g
UN Number UN2927
Beilstein 971201
Sensitivity Moisture sensitive
Solubility Information Miscible with dichloromethane,ether,terahydrofuran and protic solvents.
IUPAC Name 2,2,2-trichloroacetyl isocyanate
Formula Weight 188.4
Percent Purity 97%
Chemical Name or Material Trichloroacetyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.