accessibility menu, dialog, popup

UserName

CAS RN 302-72-7

IUPAC Name: 2-aminopropanoic acid
Synonyms: alanine alanin alanina
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: QNAYBMKLOCPYGJ-UHFFFAOYNA-N
SMILES: CC(N)C(O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)