missing translation for 'onlineSavingsMsg'
Learn More

Dl-Alanine, Technical, Fisher Chemical

Product Code. 10670131 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10670131 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670131 Supplier Fisher Chemical Supplier No. A/1160/48
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 200°C
CAS 302-72-7
pH 5.5 to 7
Color White
Molecular Formula C3H7NO2
Packaging Plastic Powder Jar
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
SMILES CC(N)C(O)=O
IUPAC Name 2-aminopropanoic acid
Molecular Weight (g/mol) 89.09
Quantity 100 g
Formula Weight 89.09g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.