accessibility menu, dialog, popup

UserName

O

CAS RN 3029-19-4

IUPAC Name: pyrene-1-carbaldehyde
Synonyms: 1-pyrenealdehyde
Molecular Weight (g/mol): 230.27
Molecular Formula: C17H10O
InChi Key: RCYFOPUXRMOLQM-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results