accessibility menu, dialog, popup

UserName

CAS RN 303186-20-1

IUPAC Name: 4-[difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-propyl-1,1'-biphenyl
Synonyms: 4-[difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-propyl-1,1'-biphenyl
Molecular Weight (g/mol): 428.35
Molecular Formula: C22H15F7O
InChi Key: YJUHXJQZJUQYJJ-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC(F)=C(C(F)=C1)C(F)(F)OC1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results