accessibility menu, dialog, popup

UserName

CAS RN 305-15-7

H2NNHClCl

CAS RN 305-15-7

IUPAC Name: (2,5-dichlorophenyl)hydrazine
Synonyms: (2,5-dichlorophenyl)hydrazine
Molecular Weight (g/mol): 177.03
Molecular Formula: C6H6Cl2N2
InChi Key: LZKWWERBNXLGLI-UHFFFAOYSA-N
SMILES: NNC1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results