accessibility menu, dialog, popup

UserName

HOBOHSN

CAS RN 305832-67-1

IUPAC Name: (5-cyanothiophen-2-yl)boronic acid
Synonyms: 5-cyanothiophen-2-ylboronic acid
Molecular Weight (g/mol): 152.96
Molecular Formula: C5H4BNO2S
InChi Key: ZEOMEPSYIIQIND-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(S1)C#N
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)