missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11369385
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369385 1 g 1g
11379385 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369385 Supplier Thermo Scientific Alfa Aesar Supplier No. L18523.03
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 305832-67-1 | Molecular Formula: C5H4BNO2S | Molecular Weight: 152.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305832-67-1
Molecular Formula C5H4BNO2S
Molecular Weight (g/mol) 152.96
MDL Number MFCD02094029
InChI Key ZEOMEPSYIIQIND-UHFFFAOYSA-N
Synonym 5-cyanothiophene-2-boronic acid, 5-cyanothiophen-2-yl boronic acid, 5-cyanothiophen-2-yl-2-boronic acid, 5-boronothiophene-2-carbonitrile, 5-cyano-2-thienyl boronic acid, 5-dihydroxyboranyl thiophene-2-carbonitrile, 5-cyano-2-thienylboronic acid, boronic acid, 5-cyano-2-thienyl, 5-cyanothiophen-2-boronic acid, acmc-1ctts
PubChem CID 4198743
IUPAC Name (5-cyanothiophen-2-yl)boronic acid
SMILES OB(O)C1=CC=C(S1)C#N

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
Formula Weight 152.97
Percent Purity 98%
Chemical Name or Material 5-Cyanothiophene-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.