accessibility menu, dialog, popup

UserName

CAS RN 3063-94-3

H3CH2COOFFFFFF

CAS RN 3063-94-3

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
Synonyms: 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 236.11
Molecular Formula: C7H6F6O2
InChi Key: FMQPBWHSNCRVQJ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results