missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stabilized, Thermo Scientific™

Product Code. 10637061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10637061 5 g Glass Bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10637061

Brand: Thermo Scientific Acros 308930050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3063-94-3
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.3000g/mL
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H6F6O2
Linear Formula H2C=C(CH3)CO2CH(CF3)2
Refractive Index 1.3300 to 1.332
MDL Number MFCD00040105
Quantity 5 g
Specific Gravity 1.3
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
Stabilizer Exact type and amount of stabilizer on the CoA
Molecular Weight (g/mol) 236.11
PubChem CID 76469
Formula Weight 236.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.