accessibility menu, dialog, popup

UserName

TiH3COH3COH3COH3CO

CAS RN 3087-36-3

IUPAC Name: titanium(4+) tetraethanolate
Synonyms: titanium ethoxide
Molecular Weight (g/mol): 228.11
Molecular Formula: C8H20O4Ti
InChi Key: JMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES: [Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (8)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)