missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide

Product Code. 10207023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207023 250 g Glass bottle 250g
10043730 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10207023 Supplier Thermo Scientific Acros Supplier No. 312542500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3087-36-3 | C8H20O4Ti | 228.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-36-3 , 67-63-0
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.15
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 150.0°C to 152.0°C (10.0 mmHg)
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 85% min
Packaging Glass bottle
Linear Formula Ti(OC2H5)4
Refractive Index 1.5033 to 1.5053
Quantity 250 g
Beilstein 01,III,1338
Specific Gravity 1.08
Solubility Information Solubility in water: hydrolysis.
Formula Weight 228.15
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material Titanium(IV) ethoxide, Contains 5-15% Isopropanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.