accessibility menu, dialog, popup

UserName

CH3OHCH3H2N

CAS RN 3096-71-7

IUPAC Name: 4-amino-2,5-dimethylphenol
Synonyms: 4-amino-2,5-dimethylphenol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: JSWVCUXQICMATE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results