accessibility menu, dialog, popup

UserName

Cl2H2H2H2H2H

CAS RN 3114-55-4

IUPAC Name: 1-chloro(²H₅)benzene
Synonyms: 1-chloro(²H₅)benzene
Molecular Weight (g/mol): 117.59
Molecular Formula: C6H5Cl
InChi Key: MVPPADPHJFYWMZ-RALIUCGRSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C([2H])=C1[2H]
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)