accessibility menu, dialog, popup

UserName

CH3ONCS

CAS RN 3125-64-2

IUPAC Name: 1-isothiocyanato-3-methoxybenzene
Synonyms: 1-isothiocyanato-3-methoxybenzene
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: WHBYCPUKGYEYFU-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)N=C=S
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results