accessibility menu, dialog, popup

UserName

NH

CAS RN 31252-42-3

IUPAC Name: 4-benzylpiperidine
Synonyms: 4-benzylpiperidine
Molecular Weight (g/mol): 175.28
Molecular Formula: C12H17N
InChi Key: ABGXADJDTPFFSZ-UHFFFAOYSA-N
SMILES: C(C1CCNCC1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)