missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperidine, 98%

Product Code. 10215880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10215880 100 g Glass bottle 100g
10743973 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215880 Supplier Thermo Scientific Acros Supplier No. 161931000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31252-42-3 | C12H17N | 175.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C(C1CCNCC1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 6.0°C to 7.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 279.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 100 g
Beilstein 20, 296
Specific Gravity 0.99
Formula Weight 175.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 4-Benzylpiperidine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.