accessibility menu, dialog, popup

UserName

CAS RN 3140-73-6

CH3ONClNOCH3N

CAS RN 3140-73-6

IUPAC Name: 2-chloro-4,6-dimethoxy-1,3,5-triazine
Synonyms: 2-chloro-4,6-dimethoxy-1,3,5-triazine
Molecular Weight (g/mol): 175.57
Molecular Formula: C5H6ClN3O2
InChi Key: GPIQOFWTZXXOOV-UHFFFAOYSA-N
SMILES: COC1=NC(OC)=NC(Cl)=N1
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)