missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-dimethoxy-1,3,5-triazine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4,6-dimethoxy-1,3,5-triazine |
| CAS | 3140-73-6 |
| Molecular Formula | C5H6ClN3O2 |
| Quantity | 5 g |
| Synonym | 4,6-Dimethoxy-2-chloro-s-triazine, 2,4-Dimethoxy-6-chloro-1,3,5-triazine, NSC 46520 |
| InChI Formula | InChI=1S/C5H6ClN3O2/c1-10-4-7-3(6)8-5(9-4)11-2/h1-2H3 |
| SMILES | COc1nc(Cl)nc(OC)n1 |
| IUPAC Name | 2-chloro-4,6-dimethoxy-1,3,5-triazine |
| Molecular Weight (g/mol) | 175.57 |
| Formula Weight | 175.0149 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction