accessibility menu, dialog, popup

UserName

CAS RN 3141-27-3

BrSBr

CAS RN 3141-27-3

IUPAC Name: 2,5-dibromothiophene
Synonyms: 2,5-dibromothiophene
Molecular Weight (g/mol): 241.93
Molecular Formula: C4H2Br2S
InChi Key: KBVDUUXRXJTAJC-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)S1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)