missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene, 95%

Product Code. 11475423
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475423 250 g 250g
11434080 25 g 25g
11465423 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475423 Supplier Thermo Scientific Alfa Aesar Supplier No. A13406.30
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3141-27-3 | C4H2Br2S | 241.93 g/mol

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

Melting Point -6°C
Density 2.15
Boiling Point 210°C to 212°C
Flash Point 99°C (210°F)
Odor Pungent
Refractive Index 1.629
Quantity 250 g
UN Number UN2810
Beilstein 109899
Solubility Information It is soluble in chloroform and methanol. It is insoluble in water.
Formula Weight 241.94
Percent Purity 96%
Chemical Name or Material 2,5-Dibromothiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.