accessibility menu, dialog, popup

UserName

CAS RN 315-13-9

CH3FNOOCH3

CAS RN 315-13-9

IUPAC Name: 1-fluoro-3,5-dimethyl-2-nitrobenzene
Synonyms: 1-fluoro-3,5-dimethyl-2-nitrobenzene
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: XGLWCYVNZDSDNQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results