accessibility menu, dialog, popup

UserName

NBrHNSNH2Br

CAS RN 31545-35-4

IUPAC Name: (3,5-dibromopyridin-2-yl)thiourea
Synonyms: 3,5-dibromopyridin-2-ylthiourea
Molecular Weight (g/mol): 311
Molecular Formula: C6H5Br2N3S
InChi Key: XIMYCSKDDBMLSY-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Br)C=C(Br)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results