accessibility menu, dialog, popup

UserName

CH3OF

CAS RN 316-68-7

IUPAC Name: 1-(4-fluoronaphthalen-1-yl)ethan-1-one
Synonyms: 1-(4-fluoronaphthalen-1-yl)ethanone
Molecular Weight (g/mol): 188.2
Molecular Formula: C12H9FO
InChi Key: GAMOBQXCYKWQLT-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2C=CC=CC2=C(F)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results