accessibility menu, dialog, popup

UserName

CAS RN 3160-40-5

H3COCl(E)

CAS RN 3160-40-5

IUPAC Name: (3E)-4-(4-chlorophenyl)but-3-en-2-one
Synonyms: 4-chlorocinnamoyl
Molecular Weight (g/mol): 180.63
Molecular Formula: C10H9ClO
InChi Key: UUKRKWJGNHNTRG-NSCUHMNNSA-N
SMILES: CC(=O)\C=C\C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)