accessibility menu, dialog, popup

UserName

CH3OPOClOH3C

CAS RN 3167-63-3

IUPAC Name: diethyl (chloromethyl)phosphonate
Synonyms: diethyl chloromethylphosphonate
Molecular Weight (g/mol): 186.57
Molecular Formula: C5H12ClO3P
InChi Key: MZBIWKMCTWJLPT-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCl)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results