missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals

Product Code. 10431402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431402 5 g Glass bottle 5g
10193011 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10431402 Supplier Thermo Scientific Acros Supplier No. 330650050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3167-63-3
Molecular Formula C5H12ClO3P
Molecular Weight (g/mol) 186.57
MDL Number MFCD00010189
InChI Key MZBIWKMCTWJLPT-UHFFFAOYSA-N
Synonym diethyl chloromethyl phosphonate, diethyl chloromethylphosphonate, diethyl chloromethanephosphonate, phosphonic acid, chloromethyl-, diethyl ester, chloromethylphosphonic acid diethyl ester, phosphonic acid, p-chloromethyl-, diethyl ester, chloromethyl phosphonic acid diethyl ester, 1-chloromethyl ethoxy phosphoryl oxyethane, acmc-1cq3s
PubChem CID 76633
IUPAC Name 1-[chloromethyl(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(CCl)OCC

Specifications

Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 109.0°C to 110.0°C (10.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Quantity 5 g
Specific Gravity 1.2
Formula Weight 186.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl (chloromethyl)phosphonate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl (chloromethyl)phosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.