accessibility menu, dialog, popup

UserName

CAS RN 31680-07-6

CH3ONOO

CAS RN 31680-07-6

IUPAC Name: 4-methyl-3-nitrobenzaldehyde
Synonyms: 4-methyl-3-nitrobenzaldehyde
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: KHWGAWBXQOKXIJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results