accessibility menu, dialog, popup

UserName

CAS RN 320-76-3

IUPAC Name: 4-bromo-2-fluoro-6-nitrophenol
Synonyms: 4-bromo-2-fluoro-6-nitrophenol
Molecular Weight (g/mol): 236
Molecular Formula: C6H3BrFNO3
InChi Key: ACQVEWFMUBXEMR-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(Br)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results