accessibility menu, dialog, popup

UserName

CH3OONOCl

CAS RN 3209-40-3

IUPAC Name: ethyl 5-(chloromethyl)-1,2-oxazole-3-carboxylate
Synonyms: ethyl 5-(chloromethyl)-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol): 189.6
Molecular Formula: C7H8ClNO3
InChi Key: RNURPQMTRDHCNM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NOC(CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results