missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(chloromethyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Product Code. 10623265
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10623265 1 g Amber Glass Bottle 1g
10226193 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623265 Supplier Thermo Scientific Maybridge Supplier No. SPB01278DA
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3209-40-3
Molecular Formula C7H8ClNO3
Molecular Weight (g/mol) 189.60
MDL Number MFCD08445409
InChI Key RNURPQMTRDHCNM-UHFFFAOYSA-N
Synonym ethyl 5-chloromethyl isoxazole-3-carboxylate, ethyl 5-chloromethyl-3-isoxazolecarboxylate, ethyl 5-chloromethyl-1,2-oxazole-3-carboxylate, ethyl 5-chloromethylisoxazole-3-carboxylate, 5-chloromethyl-isoxazole-3-carboxylic acid ethyl ester, ehyl 5-chloromethyl-1,2-oxazole-3-carboxylate, 3-isoxazolecarboxylicacid, 5-chloromethyl-, ethyl ester
PubChem CID 14674306
IUPAC Name ethyl 5-(chloromethyl)-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(CCl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.6
Percent Purity 97%
Chemical Name or Material Ethyl 5-(chloromethyl)-3-isoxazolecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.