accessibility menu, dialog, popup

UserName

CAS RN 32122-11-5

CH3OOO

CAS RN 32122-11-5

IUPAC Name: methyl 4-(benzyloxy)benzoate
Synonyms: methyl 4-(benzyloxy)benzoate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: ZLLQTDQOTFCCDF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results