missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-benzyloxybenzoate, 97%, Thermo Scientific™

Product Code. 11392686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392686 5 g 5g
11302696 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392686 Supplier Thermo Scientific Alfa Aesar Supplier No. L08749.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32122-11-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00017613
InChI Key ZLLQTDQOTFCCDF-UHFFFAOYSA-N
Synonym methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
PubChem CID 299810
IUPAC Name methyl 4-phenylmethoxybenzoate
SMILES COC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Beilstein 2119212
Formula Weight 242.27
Percent Purity 97%
Chemical Name or Material Methyl 4-benzyloxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.