accessibility menu, dialog, popup

UserName

OHHOONHOHNO

CAS RN 3237-50-1

IUPAC Name: 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
Synonyms: 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 160.09
Molecular Formula: C4H4N2O5
InChi Key: ZIIHZVKHFWOENY-UHFFFAOYSA-N
SMILES: OC1(O)C(=O)NC(=O)NC1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)