missing translation for 'onlineSavingsMsg'
Learn More

Alloxan monohydrate, 98%

Product Code. 11498216
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498216 250 g 250g
11384609 25 g 25g
11488216 50 g 50g
11394609 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498216 Supplier Thermo Scientific Alfa Aesar Supplier No. A15324.30
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3237-50-1 | C4H4N2O5 | 160.085 g/mol

Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

Solubility
Soluble in water, ethanol, acetone, glacial acetic acid and methanol. Slightly soluble in chloroform, petroleum ether, toluene, ethyl acetate and acetic anhydride. Insoluble in ether.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3237-50-1
Molecular Formula C4H4N2O5
Molecular Weight (g/mol) 160.085
MDL Number MFCD00149399
InChI Key ZIIHZVKHFWOENY-UHFFFAOYSA-N
Synonym barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6
PubChem CID 312231
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
SMILES C1(=O)C(C(=O)NC(=O)N1)(O)O

Specifications

Melting Point ∼260°C (decomposition)
Quantity 250 g
Beilstein 5309394
Merck Index 14,282
Formula Weight 160.09 (142.07 Anhydrous)
Percent Purity 98%
Chemical Name or Material Alloxan monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.