accessibility menu, dialog, popup

UserName

OHF

CAS RN 324-94-7

IUPAC Name: 4'-fluoro-[1,1'-biphenyl]-4-ol
Synonyms: 4'-fluoro-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 188.2
Molecular Formula: C12H9FO
InChi Key: QSJNKJGPJVOGPK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results