accessibility menu, dialog, popup

UserName

HOO

CAS RN 3243-42-3

IUPAC Name: 3-(naphthalen-1-yl)propanoic acid
Synonyms: 3-(naphthalen-1-yl)propanoic acid
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: PRLKVVMRQFFIOQ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results