missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)propionic Acid, TRC

Product Code. 30484003
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484003 50 mg -
30466069 25 mg 25mg
30474349 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484003 Supplier Toronto Research Chemicals Supplier No. TRCN37316350MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3243-42-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.23
IUPAC Name 3-naphthalen-1-ylpropanoic acid
SMILES OC(=O)CCc1cccc2ccccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H12O2/c14-13(15)9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7H,8-9H2,(H,14,15)
Formula Weight 200.0837
Chemical Name or Material 3-(1-Naphthyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.