accessibility menu, dialog, popup

UserName

CAS RN 3251-69-2

OHONHNHCl

CAS RN 3251-69-2

IUPAC Name: hydrogen 2-(1H-imidazol-5-yl)acetic acid chloride
Synonyms: hydrogen IMAC chloride
Molecular Weight (g/mol): 162.57
Molecular Formula: C5H7ClN2O2
InChi Key: MWHLCFYPFGFBQO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)CC1=CN=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)