missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-acetic acid monohydrochloride, 97%

Product Code. 11490647
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11490647

Brand: Thermo Scientific Alfa Aesar B22473.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3251-69-2 | C5H7ClN2O2 | 162.57 g/mol

A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3251-69-2
Molecular Formula C5H7ClN2O2
Molecular Weight (g/mol) 162.57
MDL Number MFCD00012698
InChI Key MWHLCFYPFGFBQO-UHFFFAOYSA-N
Synonym 4-imidazoleacetic acid hydrochloride, 2-1h-imidazol-5-yl acetic acid hydrochloride, imidazole-4-acetic acid hydrochloride, 2-1h-imidazol-4-yl acetic acid hydrochloride, 4-imidazoleacetic acid hcl, imidazoleacetic acid hydrochloride, unii-et7a74roao, 4-imidazoleaceticacidhydrochloride, i4aa, et7a74roao
PubChem CID 145685
IUPAC Name 2-(1H-imidazol-5-yl)acetic acid;hydrochloride
SMILES Cl.OC(=O)CC1=CN=CN1

Specifications

Melting Point 221°C to 224°C
Odor Odorless
Quantity 1 g
Beilstein 3701591
Solubility Information Soluble in water.
Formula Weight 162.58
Percent Purity 97%
Chemical Name or Material Imidazole-4-acetic acid monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.