accessibility menu, dialog, popup

UserName

CH3ONNCl

CAS RN 325685-59-4

IUPAC Name: 4-chloro-6-(methoxymethyl)-2-phenylpyrimidine
Synonyms: 4-chloro-6-(methoxymethyl)-2-phenylpyrimidine
Molecular Weight (g/mol): 234.68
Molecular Formula: C12H11ClN2O
InChi Key: GSSSCZAOXFZADM-UHFFFAOYSA-N
SMILES: COCC1=CC(Cl)=NC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results