accessibility menu, dialog, popup

UserName

FN

CAS RN 326-62-5

IUPAC Name: 2-(2-fluorophenyl)acetonitrile
Synonyms: benzeneacetonitrile, 2-fluoro-
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: DAVJMKMVLKOQQC-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CC#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results