accessibility menu, dialog, popup

UserName

CAS RN 3262-72-4

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C8H15NO5
InChi Key: FHOAKXBXYSJBGX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (6)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (6)
  • (2)
  • (3)