missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-serine, 98% (dry wt.), may cont. up to 10% water

Product Code. 11493327
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493327 25 g 25g
11473327 1 g 1g
11483327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493327 Supplier Thermo Scientific Alfa Aesar Supplier No. A16224.14
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3262-72-4 | C8H15NO5 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3262-72-4
Molecular Formula C8H15NO5
Molecular Weight (g/mol) 205.21
MDL Number MFCD00037243
InChI Key FHOAKXBXYSJBGX-MLHKIVSYNA-N
Synonym boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser
PubChem CID 98766
SMILES CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O

Specifications

Melting Point 90°C to 91°C (decomposition)
Quantity 25 g
Beilstein 2212252
Optical Rotation −3.5° (c=2 in Acetic Acid)
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Formula Weight 205.21
Percent Purity 98%
Assay (dry wt.), may cont. up to 10% water
Chemical Name or Material N-Boc-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.