accessibility menu, dialog, popup

UserName

CAS RN 32815-70-6

H3CCH3OHCH2

CAS RN 32815-70-6

IUPAC Name: 2-methylhex-5-en-3-ol
Synonyms: 2-methylhex-5-en-3-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: JKGMJZOZIJHPOH-UHFFFAOYNA-N
SMILES: CC(C)C(O)CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results