accessibility menu, dialog, popup

UserName

BClFHOOH

CAS RN 328956-61-2

IUPAC Name: (3-chloro-5-fluorophenyl)boronic acid
Synonyms: 3-chloro-5-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: XYQDHVBKUNNLGZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results