accessibility menu, dialog, popup

UserName

CAS RN 329-49-7

FHOBrNOO

CAS RN 329-49-7

IUPAC Name: 2-bromo-6-fluoro-4-nitrophenol
Synonyms: 2-bromo-6-fluoro-4-nitrophenol
Molecular Weight (g/mol): 236
Molecular Formula: C6H3BrFNO3
InChi Key: YJBBEHQOMCJIGN-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=C1Br)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results