accessibility menu, dialog, popup

UserName

CAS RN 3290-06-0

ClClCl

CAS RN 3290-06-0

IUPAC Name: 1,3-dichloro-5-(chloromethyl)benzene
Synonyms: 3,5-dichlorobenzyl chloride
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: ZFLRKAMKGYNFPH-UHFFFAOYSA-N
SMILES: ClCC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results